Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124991.4 | NP_200420.1 | 5 - 6 | 1228 | 118 | |
GenBank mRNA | AK117496.1 | - | 5 - 6 | 1103 | 118 | |
BT004976.1 | - | 5 - 6 | 1065 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |