Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203996.1 | NP_001190925.1 | 8 - 9 | 1180 | 75 | |
NM_119710.5 | NP_195270.1 | 8 - 9 | 1118 | 75 | ||
NM_179166.1 | NP_849497.1 | 7 - 8 | 1099 | 75 | ||
NM_179167.2 | NP_849498.1 | 8 - 9 | 1137 | 75 | ||
NM_179168.1 | NP_849499.1 | 6 - 7 | 954 | 75 | ||
GenBank mRNA | AF386982.1 | - | 7 - 8 | 1099 | 75 | |
AK316831.1 | - | 8 - 9 | 1006 | 75 | ||
AK317167.1 | - | 8 - 9 | 998 | 75 | ||
AY081477.1 | - | 7 - 8 | 924 | 75 | ||
AY087377.1 | - | 8 - 9 | 1054 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |