Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IAA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100334.4 | NP_171949.1 | 1 - 2 | 492 | 76 | |
NM_179258.2 | NP_849589.1 | 1 - 2 | 411 | 76 | ||
GenBank mRNA | AF332398.1 | - | 1 - 2 | 204 | 76 | |
AY065240.1 | - | 1 - 2 | 346 | 76 | ||
AY096535.1 | - | 1 - 2 | 204 | 76 | ||
U18414.1 | - | 1 - 2 | 303 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |