Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331836.1 | NP_001318964.1 | 4 - 5 | 1370 | 111 | |
NM_001331837.1 | NP_001318965.1 | 4 - 5 | 1430 | 111 | ||
NM_001331838.1 | NP_001322700.1 | 3 - 4 | 1794 | 111 | ||
NM_001331839.1 | NP_001322701.1 | 2 - 3 | 1767 | 111 | ||
NM_100824.3 | NP_172424.1 | 3 - 4 | 1343 | 111 | ||
NM_179295.3 | NP_849626.1 | 3 - 4 | 1403 | 111 | ||
GenBank mRNA | AF100166.1 | - | 3 - 4 | 1035 | 111 | |
AF251693.1 | - | 2 - 3 | 952 | 111 | ||
AK117255.1 | - | 3 - 4 | 1088 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |