Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAT-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331880.1 | NP_001320930.1 | 1 - 2 | 435 | 79 | |
NM_001331881.1 | NP_001320929.1 | 1 - 2 | 435 | 79 | ||
NM_001331882.1 | NP_001320928.1 | 1 - 2 | 398 | 79 | ||
NM_001331883.1 | NP_001320927.1 | 1 - 2 | 269 | 79 | ||
NM_100880.3 | NP_563859.1 | 1 - 2 | 269 | 79 | ||
NM_179298.2 | NP_849629.1 | 1 - 2 | 269 | 79 | ||
GenBank mRNA | AJ276123.1 | - | 1 - 2 | 149 | 79 | |
AK316768.1 | - | 1 - 2 | 198 | 79 | ||
AY054517.1 | - | 1 - 2 | 270 | 79 | ||
AY128778.1 | - | 1 - 2 | 63 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |