Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP45B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101037.4 | NP_172630.1 | 2 - 3 | 841 | 60 | |
NM_179310.2 | NP_849641.1 | 2 - 3 | 841 | 60 | ||
GenBank mRNA | AY093201.1 | - | 2 - 3 | 812 | 60 | |
BT008494.1 | - | 2 - 3 | 720 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Arabidopsis thaliana Probe spans exons |