Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112163.6 | NP_187929.4 | 8 - 9 | 1548 | 89 | |
GenBank mRNA | AY063883.1 | - | 8 - 9 | 1507 | 89 | |
AY142588.1 | - | 8 - 9 | 1476 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |