Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112177.3 | NP_566452.1 | 10 - 11 | 1568 | 68 | |
GenBank mRNA | AF424628.1 | - | 10 - 11 | 1253 | 68 | |
AY097343.1 | - | 9 - 10 | 1104 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |