Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSP60-3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112240.4 | NP_566466.1 | 3 - 4 | 395 | 73 | |
GenBank mRNA | AY094453.1 | - | 3 - 4 | 309 | 73 | |
BT015923.1 | - | 3 - 4 | 193 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |