Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APG12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338094.1 | NP_001326256.1 | 1 - 2 | 99 | 66 | |
NM_001338095.1 | NP_001326257.1 | 1 - 2 | 90 | 66 | ||
NM_001338096.1 | NP_001326255.1 | 1 - 2 | 90 | 66 | ||
NM_112251.4 | NP_188013.2 | 1 - 2 | 99 | 66 | ||
GenBank mRNA | AB073185.1 | - | 1 - 2 | 43 | 66 | |
AK118997.1 | - | 1 - 2 | 90 | 66 | ||
BT024653.1 | - | 1 - 2 | 36 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |