Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338114.1 | NP_001326872.1 | 4 - 5 | 722 | 83 | |
NM_112278.3 | NP_188037.1 | 3 - 4 | 1064 | 83 | ||
GenBank mRNA | AK221834.1 | - | 3 - 4 | 640 | 83 | |
AY054230.1 | - | 3 - 4 | 618 | 83 | ||
AY062690.1 | - | 3 - 4 | 620 | 83 | ||
BT002522.1 | - | 3 - 4 | 620 | 83 | ||
BT008484.1 | - | 3 - 4 | 577 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |