Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL04 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202964.2 | NP_001189893.1 | 14 - 15 | 1815 | 98 | |
NM_001338124.1 | NP_001326811.1 | 14 - 15 | 1935 | 98 | ||
NM_112317.3 | NP_188075.2 | 14 - 15 | 1974 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |