Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL04 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202964.2 | NP_001189893.1 | 37 - 38 | 4322 | 64 | |
NM_001338124.1 | NP_001326811.1 | 36 - 37 | 4472 | 64 | ||
NM_112317.3 | NP_188075.2 | 36 - 37 | 4559 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |