Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338143.1 | NP_001326245.1 | 9 - 10 | 3633 | 117 | |
NM_001338145.1 | NP_001326243.1 | 7 - 8 | 3752 | 117 | ||
NM_001338146.1 | NP_001326244.1 | 9 - 10 | 3423 | 117 | ||
NM_001338147.1 | NP_001326242.1 | 8 - 9 | 3196 | 117 | ||
NM_112360.3 | NP_188116.1 | 8 - 9 | 3563 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |