Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342124.1 | NP_001329752.1 | 5 - 6 | 772 | 76 | |
NM_119353.4 | NP_194929.2 | 7 - 8 | 1120 | 76 | ||
GenBank mRNA | AB206554.1 | - | 3 - 4 | 306 | 76 | |
AK228013.1 | - | 7 - 8 | 975 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |