Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203962.1 | NP_001190891.1 | 3 - 4 | 617 | 107 | |
NM_119407.3 | NP_567896.1 | 3 - 4 | 673 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |