Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PME44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342204.1 | NP_001328318.1 | 3 - 4 | 2475 | 86 | |
NM_119476.7 | NP_567917.4 | 3 - 4 | 1117 | 86 | ||
GenBank mRNA | AF378892.1 | - | 3 - 4 | 946 | 86 | |
BT029770.1 | - | 2 - 3 | 517 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |