Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203975.1 | NP_001190904.1 | 1 - 2 | 578 | 108 | |
NM_119497.5 | NP_567920.1 | 1 - 2 | 633 | 108 | ||
GenBank mRNA | FJ708762.1 | - | 1 - 2 | 89 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |