Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119573.5 | NP_567952.1 | 1 - 2 | 496 | 66 | |
GenBank mRNA | AF360331.1 | - | 1 - 2 | 264 | 66 | |
AY056339.1 | - | 1 - 2 | 209 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |