Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCD3;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119579.3 | NP_195142.1 | 3 - 4 | 1292 | 77 | |
GenBank mRNA | AK221712.1 | - | 3 - 4 | 984 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |