Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179694.4 | NP_850025.1 | 7 - 8 | 1260 | 65 | |
GenBank mRNA | AY070402.1 | - | 7 - 8 | 928 | 65 | |
AY096578.1 | - | 7 - 8 | 873 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |