Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336021.1 | NP_001323723.1 | 5 - 6 | 684 | 86 | |
NM_179739.4 | NP_850070.1 | 5 - 6 | 840 | 86 | ||
NM_201804.3 | NP_973533.1 | 5 - 6 | 684 | 86 | ||
NM_201805.1 | NP_973534.1 | 5 - 6 | 591 | 86 | ||
GenBank mRNA | AY081299.1 | - | 5 - 6 | 627 | 86 | |
BT001221.1 | - | 5 - 6 | 608 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |