Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIPK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036350.1 | NP_001031427.1 | 1 - 2 | 286 | 158 | |
NM_001336096.1 | NP_001318295.1 | 1 - 2 | 292 | 158 | ||
NM_179761.2 | NP_850092.1 | 1 - 2 | 286 | 158 | ||
NM_179763.4 | NP_850094.1 | 2 - 3 | 391 | 158 | ||
NM_179764.3 | NP_850095.2 | 2 - 3 | 464 | 158 | ||
GenBank mRNA | AF367290.1 | - | 1 - 2 | 286 | 158 | |
AY059163.1 | - | 1 - 2 | 168 | 158 | ||
AY091098.1 | - | 1 - 2 | 279 | 158 | ||
AY142671.1 | - | 1 - 2 | 168 | 158 | ||
AY266298.1 | - | 1 - 2 | 168 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |