Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036358.2 | NP_001031435.1 | 3 - 4 | 810 | 90 | |
NM_001336153.1 | NP_001323735.1 | 3 - 4 | 543 | 90 | ||
NM_001336154.1 | NP_001323736.1 | 3 - 4 | 284 | 90 | ||
NM_001336155.1 | NP_001323737.1 | 3 - 4 | 669 | 90 | ||
NM_001336156.1 | NP_001323738.1 | 3 - 4 | 592 | 90 | ||
NM_179784.2 | NP_850115.1 | 2 - 3 | 779 | 90 | ||
GenBank mRNA | AY065457.1 | - | 2 - 3 | 331 | 90 | |
BT004417.1 | - | 2 - 3 | 161 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |