Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202971.2 | NP_001189900.1 | 4 - 5 | 555 | 87 | |
NM_001338166.1 | NP_001327652.1 | 5 - 6 | 1164 | 87 | ||
NM_112407.5 | NP_188158.4 | 6 - 7 | 1209 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |