Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UVR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035626.2 | NP_001030703.1 | 11 - 12 | 1611 | 130 | |
NM_112432.2 | NP_566520.1 | 12 - 13 | 1652 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |