Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIFI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035630.1 | NP_001030707.1 | 3 - 4 | 350 | 85 | |
NM_001125170.1 | NP_001118642.1 | 2 - 3 | 371 | 85 | ||
NM_001202975.1 | NP_001189904.1 | 3 - 4 | 460 | 85 | ||
NM_112454.3 | NP_566528.1 | 3 - 4 | 447 | 85 | ||
GenBank mRNA | AY035025.1 | - | 3 - 4 | 306 | 85 | |
AY133879.1 | - | 3 - 4 | 269 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |