Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338233.1 | NP_001319568.1 | 2 - 3 | 263 | 104 | |
NM_001338234.1 | NP_001326890.1 | 3 - 4 | 654 | 104 | ||
NM_112518.5 | NP_566547.2 | 2 - 3 | 258 | 104 | ||
GenBank mRNA | AY059903.1 | - | 2 - 3 | 243 | 104 | |
AY114681.1 | - | 1 - 2 | 193 | 104 |
Gene Symbol: AT4G01610 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK230235.1 | - | 1044 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |