Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112527.2 | NP_566552.2 | 3 - 4 | 747 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |