Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084707.2 | NP_001078176.1 | 8 - 9 | 1180 | 140 | |
NM_001338322.1 | NP_001325994.1 | 11 - 12 | 1420 | 140 | ||
NM_001338323.1 | NP_001325998.1 | 10 - 11 | 1379 | 140 | ||
NM_001338324.1 | NP_001325997.1 | 7 - 8 | 1247 | 140 | ||
NM_001338325.1 | NP_001325995.1 | 6 - 7 | 1015 | 140 | ||
NM_001338326.1 | NP_001325996.1 | 7 - 8 | 948 | 140 | ||
NM_112689.2 | NP_188435.2 | 11 - 12 | 1411 | 140 | ||
GenBank mRNA | DQ446670.1 | - | 8 - 9 | 1180 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |