Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338385.1 | NP_001327751.1 | 9 - 10 | 5307 | 95 | |
NM_001338386.1 | NP_001327752.1 | 9 - 10 | 5307 | 95 | ||
NM_112806.3 | NP_188550.3 | 9 - 10 | 5055 | 95 | ||
GenBank mRNA | HQ446214.1 | - | 9 - 10 | 5067 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |