Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112947.4 | NP_188691.1 | 5 - 6 | 721 | 102 | |
GenBank mRNA | AY062626.1 | - | 5 - 6 | 638 | 102 | |
BT000005.1 | - | 4 - 5 | 538 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |