Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119684.4 | NP_567977.2 | 4 - 5 | 1169 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |