Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342391.1 | NP_001328787.1 | 4 - 5 | 397 | 55 | |
NM_119763.3 | NP_195321.1 | 4 - 5 | 417 | 55 | ||
GenBank mRNA | AK119023.1 | - | 4 - 5 | 397 | 55 | |
BT006053.1 | - | 4 - 5 | 352 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |