Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204003.1 | NP_001190932.1 | 9 - 10 | 1232 | 110 | |
NM_119774.3 | NP_195331.2 | 9 - 10 | 1311 | 110 | ||
GenBank mRNA | DQ446901.1 | - | 9 - 10 | 1200 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |