Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036719.2 | NP_001031796.1 | 8 - 9 | 971 | 87 | |
NM_001342410.1 | NP_001329288.1 | 8 - 9 | 1241 | 87 | ||
NM_119811.3 | NP_568005.1 | 8 - 9 | 1241 | 87 | ||
GenBank mRNA | AB063254.1 | - | 7 - 8 | 783 | 87 | |
AY120759.1 | - | 8 - 9 | 1059 | 87 | ||
BT000131.1 | - | 7 - 8 | 762 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |