Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342416.1 | NP_001328936.1 | 3 - 4 | 364 | 97 | |
NM_001342417.1 | NP_001328935.1 | 3 - 4 | 364 | 97 | ||
NM_119840.7 | NP_195394.4 | 3 - 4 | 444 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |