Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204009.1 | NP_001190938.1 | 6 - 7 | 847 | 68 | |
NM_119856.3 | NP_195410.1 | 5 - 6 | 1006 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Inventoried Probe spans exons |