Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRFR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342443.1 | NP_001329700.1 | 16 - 17 | 2776 | 116 | |
NM_119910.4 | NP_195462.3 | 16 - 17 | 2776 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |