Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSLB03 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179859.4 | NP_850190.1 | 7 - 8 | 1617 | 79 | |
GenBank mRNA | AY090243.1 | - | 7 - 8 | 1408 | 79 | |
BT002290.1 | - | 7 - 8 | 1353 | 79 | ||
KJ138881.1 | - | 7 - 8 | 1353 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |