Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179881.4 | NP_850212.1 | 7 - 8 | 1168 | 105 | |
GenBank mRNA | AY074317.1 | - | 7 - 8 | 1051 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |