Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRFL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036420.2 | NP_001031497.1 | 4 - 5 | 1230 | 82 | |
NM_001336621.1 | NP_001324498.1 | 5 - 6 | 528 | 82 | ||
NM_179944.3 | NP_850275.1 | 5 - 6 | 606 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |