Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RA-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100093.4 | NP_171715.1 | 7 - 8 | 653 | 69 | |
GenBank mRNA | AY052333.1 | - | 7 - 8 | 577 | 69 | |
AY061905.1 | - | 6 - 7 | 522 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |