Search
Search

CLPP5| Gene Symbol: CLPP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100137.4 | NP_563657.1 | 1 - 2 | 388 | 96 | |
| GenBank mRNA | AB022326.1 | - | 1 - 2 | 181 | 96 | |
| AJ012278.1 | - | 1 - 2 | 151 | 96 | ||
| AY084394.1 | - | 1 - 2 | 229 | 96 | ||
| BT024859.1 | - | 1 - 2 | 128 | 96 | ||
Set Membership: |
|