Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100137.4 | NP_563657.1 | 6 - 7 | 838 | 67 | |
GenBank mRNA | AB022326.1 | - | 6 - 7 | 631 | 67 | |
AJ012278.1 | - | 6 - 7 | 601 | 67 | ||
AY084394.1 | - | 6 - 7 | 679 | 67 | ||
BT024859.1 | - | 6 - 7 | 578 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |