Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRP39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197976.1 | NP_001184905.1 | 4 - 5 | 784 | 122 | |
NM_001331460.1 | NP_001322270.1 | 3 - 4 | 839 | 122 | ||
NM_100289.4 | NP_563700.1 | 4 - 5 | 753 | 122 | ||
GenBank mRNA | AY056321.1 | - | 4 - 5 | 718 | 122 | |
AY087878.1 | - | 4 - 5 | 720 | 122 | ||
AY091187.1 | - | 3 - 4 | 570 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |