Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRP39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160830.2 | NP_001154302.1 | 2 - 3 | 603 | 99 | |
NM_001197976.1 | NP_001184905.1 | 5 - 6 | 957 | 99 | ||
NM_001331460.1 | NP_001322270.1 | 4 - 5 | 1012 | 99 | ||
NM_001331461.1 | NP_001322271.1 | 2 - 3 | 423 | 99 | ||
NM_100289.4 | NP_563700.1 | 5 - 6 | 926 | 99 | ||
GenBank mRNA | AY056321.1 | - | 5 - 6 | 891 | 99 | |
AY087878.1 | - | 5 - 6 | 893 | 99 | ||
AY091187.1 | - | 4 - 5 | 743 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |