Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197977.1 | NP_001184906.1 | 7 - 8 | 4066 | 151 | |
NM_001331463.1 | NP_001322077.1 | 8 - 9 | 3933 | 151 | ||
NM_100293.3 | NP_171908.1 | 7 - 8 | 4099 | 151 | ||
GenBank mRNA | AF225908.1 | - | 7 - 8 | 3834 | 151 | |
Y11250.1 | - | 3814 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |