Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100331.4 | NP_171946.2 | 1 - 2 | 820 | 86 | |
GenBank mRNA | BT010866.1 | - | 1 - 2 | 484 | 86 | |
BT011789.1 | - | 1 - 2 | 420 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |