Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPS3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100370.3 | NP_171984.2 | 10 - 11 | 3108 | 58 | |
GenBank mRNA | AY078949.1 | - | 10 - 11 | 3019 | 58 | |
BT002210.1 | - | 10 - 11 | 2911 | 58 | ||
KJ139031.1 | - | 10 - 11 | 2911 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |